missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Temperature/Humidity Meter

Simultaneously displays humidity, dew point and temperature

Brand:  Fisherbrand™ 90029-53

Product Code. 11789825


Description

Description

Simultaneously displays humidity, dew point and temperature. Fisher Scientific™ Traceable™ Temperature/Humidity Meter are ideal for use in cleanrooms, food-processing facilities, laboratories, and other humidity-sensitive areas.

  • Serial RS-232 output allows connection to a computer to download datalogger results for constant monitoring
  • Datalogger captures 1000 readings manually or automatically when set between 1 and 3600 second (sold separately)
  • Temperature range, 0° to 50°C (32° to 122°F), with 0.01° resolution, ±1°C accuracy
  • Relative humidity range, 10 to 95% RH, with 0.01% resolution, ±3% accuracy
  • Memory recalls maximum, minimum, and average readings
  • HOLD button freezes display for later reading
  • Large display (1.37 in.H) shows temperature and humidity simultaneously

Order Info

Measures 1.25W x 3L x 7 in. H (3.2 x 7.6 x 17.8cm); weighs 9.5 oz. (269g)

Specifications
Show More
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Traceable™ Temperature/Humidity Meter > Temperature/humidity meter

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.