Learn More
Fisherbrand™ Vortex Mixer Accessories: Foam Insert Kit
For use with Fisherbrand vortex mixers.
Brand:  Fisherbrand™ 80863002
 View more versions of this product
Product Code. 15786949
  • 28.05 € / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
Foam Insert Variety Pack
  • Includes foam inserts for the following: 9 to 13mm tube holder, 14 to 19mm tube holder, 20 to 29mm
Specifications
Specifications
Blank Foam insert
Foam
Vortex mixer