Learn More
Fisherbrand™ Vortex Mixer Accessories: Head and Head Cover
For use with Fisherbrand vortex mixers.
Brand:  Fisherbrand™ 80863007
Product Code. 15736959
  • 66.00 € / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For use with Fisherbrand vortex mixers.
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
  • 3 in. Head permits easy-touch mixing of beakers and flasks
  • Head Cover slides over top of head—ideal for odd-shaped containers
  • Head With Rubber Cover is designed for mixing irregular shaped objects
Specifications
Specifications
Head with Cover
76 mm
Fisherbrand Vortex mixer
Head, Rubber cover