missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Wellwash™ and Wellwash™ Versa™ Microplate Strip Washer: Spare Bottles

Accessories for Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer

Brand:  Thermo Scientific™ N10822

 View more versions of this product

Product Code. 11717996

  • 217.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

For use with Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer

Subsitute bottles of different sizes for added convenience

ELISA-based assays, cell washing

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Solid cap
4 L
Storage Bottle
135.25 oz.
Round
Wellwash Microplate Strip Washer, Wellwash Versa Microplate Strip Washer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers