missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ FB-VE10-1 Vertical Electrophoresis System: Clamps

For use with FB-VE10-1 vertical electrophoresis systems

Brand:  Fisherbrand™ P8-CL

Product Code. 11923286

  • 57.95 € / Pack of 2
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Set of 2; 10 x 10cm.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

FisherBiotech FB-VE10-1 Vertical Electrophoresis System; Owl™ P8DS Gel System
Side Clamp
10 cm
10 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers