Learn More
Fisherbrand™ 3/8 in. Dot Labels
GREENER_CHOICE
Fisher branded Cryogenic Storage Labels use a special adhesive that is designed to withstand extreme cold temperatures as low as -196º C (-311º F). Our cryogenic storage labels can be printed or written on and will not bleed, smudge or smear. Engineered for you to use in your laser printer.
Brand:  Fisherbrand™ 11010201
Product Code. 17837191
  • 58.00 € / 20 sheets
Estimated Shipment: 11-07-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Blue
Non-sterile
192 dots/sheet; 20 sheets/Pk.
9.55 mm
Dot label laser sheet
0.5 to 1.5mL tubes
0.37 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ 3/8 in. Dot Labels > Sheet; Blue