missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ 384-Well ShallowWell Polypropylene Microplates
Small volume applications
609.63 € - 839.00 €
Specifications
Certifications/Compliance | ANSI |
---|---|
Format | Standard |
Array | 16 x 24 |
Material | Polypropylene |
No. of Wells | 384 |
Product Code | Brand | Volume (Metric) Well | Sterility | Color | For Use With (Application) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) Well | Sterility | Color | For Use With (Application) | Price | Quantity & Availability | ||
13565450
|
Fisherbrand™
267457 |
120 μL | Non-sterile | White | Storage or colorimetric assays |
632.00 €
Pack of 100 |
Estimated Shipment: 25-03-2025
Log in to see stock. |
||
13575450
|
Fisherbrand™
267463 |
58 μL | Sterile | White | Storage or colorimetric assays |
839.00 €
Pack of 100 |
Estimated Shipment: 25-03-2025
Log in to see stock. |
||
13585450
|
Fisherbrand™
267465 |
120 μL | Non-sterile | White | Luminescent assays | N/A | |||
13595450
|
Fisherbrand™
267464 |
120 μL | Non-sterile | Black | Fluorescent assays |
748.00 €
Pack of 100 |
In Stock
Log in to see stock. |
||
Description
Feature a shared-wall design which yields increased well volume
- Automation compatible, ANSI standard
- Low-binding and chemically resistant surface
- Natural plates for storage or colorimetric assays
- White plates for luminescent assays, including SPA
- Black plates for fluorescent assays
- Round bottom wells for optimal sample recovery
- Rounded square wells minimize wicking (capillary action)
- Working volume range: 2 to 35μL/well
Order Info
Lids not included
Specifications
ANSI | |
16 x 24 | |
384 |
Standard | |
Polypropylene | |
Round, Conical Bottom (V-well) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title