missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Absorbent Underpads
GREENER_CHOICE

Quickly absorb liquids and keep spills from spreading

Brand:  Fisherbrand™ 55100-00

 View more versions of this product

Product Code. 11977964

  • 171.00 € / Pack of 400
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

Absorb liquids quickly and contain spills

Cushioned surface minimizes breakage of delicate labware

  • Made with high-absorbency fibers
  • Polyethylene backing is chemical resistant, and the spunbond top layer provides durability even when wet
  • Lie flat, hugging the benchtop to prevent sliding
  • Absorb up to 750mL/m2

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Absorbent Underpad
750 mL/m2
30 m
510 mm
Quickly absorb liquids and keep spills from spreading
Polyethylene
100 ft.
20 in.
100 ft. x 20 in. (30m x 510mm)
Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.