missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Absorption Macro Rectangular Cells with PTFE Lid
Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
300 μL
350 μL
1500 μL
1750 μL
3000 μL
Material:
Optical Glass
Optical Quartz
Quartz
Special Optical Glass
Path Length:
1 mm
5 mm
10 mm
Quantity:
2/Pk., 4 Pk./Cs.
2/Pk., 8 Pk./Cs.
4/Pk., 4 Pk./Cs.
4/Pk., 8 Pk./Cs.
Ea., 2 Ea./Cs.
Ea., 8 Ea./Cs.
missing translation for 'unitSize'
Each
Pack of 2
Pack of 4
Description
Includes
PTFE lid
Fit standard spectrophotometer cell holders
- All cells are 12.5mm wide
- 1, 5 or 10mm path length rectangular cells
- Available with a variety of window materials for wide range of assays
- Available in four materials e.g. Quartz, Optical Quartz, Special Optical Glass and Optical Glass
- Quartz windows for transmission from far ultraviolet through the infrared
- Glass windows for routine work in ultraviolet region and visible region
Specifications
Specifications
| Type | Macro Absorption Cell |
| Capacity (Metric) | 3000 μL |
| Material | Special Optical Glass |
| Closure Material | PTFE |
| Width (Metric) Interior | 10mm |
| Quantity | 2/Pk., 8 Pk./Cs. |
| Path Length | 10 mm |
| Wavelength Range | 320 to 2500 nm |
| Height (Metric) | 45 mm |
| For Use With (Equipment) | Standard Spectrophotometer Cell Holder |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction