missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Absorption Quartz Semi Micro Rectangular Cells with PTFE Lid, w/PTFE lid
Fit standard spectrophotometer cell holders
402.00 € - 825.00 €
Specifications
Type | Semi-micro Absorption Cell |
---|---|
Capacity (Metric) | 1000 μL |
Material | Quartz |
Closure Material | PTFE |
Width (Metric) Interior | 4mm |
Includes: PTFE lid
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15246033
|
Fisherbrand™
14958127 |
4/Pk., 8 Pk./Cs. |
825.00 €
Pack of 4 |
Estimated Shipment: 11-06-2025
Log in to see stock. |
|||||
15226033
|
Fisherbrand™
14958125 |
2/Pk., 8 Pk./Cs. |
402.00 €
Pack of 2 |
Estimated Shipment: 19-02-2025
Log in to see stock. |
|||||
Description
Fit standard spectrophotometer cell holders
- All cells are 12.5mm wide
- 10mm path length rectangular cells
- Reduce reagent use with Semimicro cells
- Quartz windows for transmission from far ultraviolet through the infrared
Specifications
Semi-micro Absorption Cell | |
Quartz | |
4mm | |
200 to 2500 nm | |
Standard Spectrophotometer Cell Holder | |
Transmission from far ultraviolet through the infrared |
1000 μL | |
PTFE | |
10 mm | |
45 mm | |
12.5 mm | |
Rectangle |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title