Learn More
Fisherbrand™ Aluminum Lab Frame Rods
Corrosion resistant hard aluminum rods for smooth fit with lab frames.
Brand:  Fisherbrand™ 02215478
 View more versions of this product
Product Code. 17291384
  • 14.00 € / Each
Explore more special offers
Description
Description
  • Rods with center-less ground allow for precise fiting
  • With chamfered ends for easy location
  • Diameter: 13mm (0.51 in.)
Specifications
Specifications
Aluminum
12 in.
30.48 cm
Corrosion resistant with chamfered ends for easy location