missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Aluminum Dissecting Pan

Fisherbrand Aluminum Dissecting Pan is a major tool used in dissection across the globe.

Brand:  Fisherbrand™ 1381461

 View more versions of this product

Product Code. 13403746

  • 24.00 € / Each
Estimated Shipment: 16-06-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Fisherbrand™ Aluminum Dissecting Pan is a world renown dissection tool designed for all a variety of dissection experiments
  • Major tool in dissection experiments
  • Made to last and feature safe, smooth, rolled edges for lasting rigidity and durability
  • Stack for compact storage
  • Rustproof
  • Available with or without black dissecting wax
  • Used in the fields of education and research
  • Made from ASTM™ standard aluminum

Order Info

Dimension: 11 x 7 x 1.5 in.

Specifications

Specifications

11 in. x 7 in. x 1.5 in.
Wax
1 Ea.
General dissecting use
Aluminum
Aluminum Dissecting Pan
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Aluminum Dissecting Pan > With Wax

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.