Learn More
Fisherbrand™ Aluminum Dissecting Pan
Fisherbrand Aluminum Dissecting Pan is a major tool used in dissection across the globe.
Brand:  Fisherbrand™ 1381460
 View more versions of this product
Product Code. 17199180
  • 15.00 € / Each
Estimated Shipment: 19-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Fisherbrand™ Aluminum Dissecting Pan is a world renown dissection tool designed for all a variety of dissection experiments
  • Major tool in dissection experiments
  • Made to last and feature safe, smooth, rolled edges for lasting rigidity and durability
  • Stack for compact storage
  • Rustproof
  • Available with or without black dissecting wax
  • Used in the fields of education and research
  • Made from ASTM™ standard aluminum
Order Info
Dimension: 11 x 7 x 1.5 in.
Specifications
Specifications
General dissecting use
Aluminum
Aluminum Dissecting Pan
Without wax
1 Ea.