missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Basic Protection Disposable Pleated Bouffant Caps

Product Code. 15451942
Click to view available options
Color:
Blue
White
Size:
21 in.
24 in.
28 in.
This item is not returnable. View return policy
This item is not returnable. View return policy

Bouffant features an encapslated elastic headband in soft poly thread for worker comfort and secure protection.

  • Disposable
  • Pleated
  • Polypropylene
  • Choice of white or blue

Specifications

Product Type Bouffant
Size 24 in.
Material Spunbond Polypropylene
Color White
Disposable Single-use
For Use With (Application) Laboratory Protection
Style Pleated
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'