missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Basic Protection Polypropylene Disposable Lab Coats with Elastic Cuffs
Latex-free spunbond polypropylene construction lab ware
Specifications
| Color | White |
|---|---|
| Material | Polypropylene |
| Closure Type | Snaps |
| Cuff Style | Elastic |
| No. of Pockets | None |
Description
Latex-free spunbond polypropylene construction lab ware
- Single-use lab coat
- Standard collar, no pockets
- Packed 25 lab coats/case
- Elastic cuffs hold sleeves in place
- Five snaps for a secure closure
- Fabric weight: 50g/m2
- White
Specifications
| White | |
| Snaps | |
| None | |
| Yes |
| Polypropylene | |
| Elastic | |
| Standard |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title