Learn More
Fisherbrand™ Bitran™ Specimen Storage Bags
Choice of Saranex™ odor-control film for long-term storage or polyethylene for short-term storage
186.00 €
Specifications
Closure Type Zip Seal
View More Specs
Products 1
Product Code Brand Dimensions (L x W) Material Length (Metric) Width (Metric) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Material Length (Metric) Width (Metric) Price Quantity & Availability  
16488115
View Documents
Fisherbrand™
19240093
2 x 4 in. (5.1 x 10.2cm) Saranex Film 5.1 cm 10.2 cm
186.00 €
Pack of 250
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
Description
Description
Choice of Saranex™ odor-control film for long-term storage or polyethylene for short-term storage
  • Use for collecting, transporting and storing tissue specimens
  • Minimize storage space and help to eliminate leakage, breakage and contamination
  • Resealable double-lock zipper closure
  • Designed for storage of tissues and fixatives and for use as mini-anaerobic environments
Specifications
Specifications
Zip Seal
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Bitran™ Specimen Storage Bags