missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Blood Collection Tray
Designed with phlebotomists in mind. Fisherbrand™ Blood Collection Tray was developed after extensive research in clinics and hospitals. This lightweight solution for organizing and transporting tubes and smaller items includes a sturdy plastic tray, 36-well tube rack and disposable inserts.
Brand: Fisherbrand™ HS2200AF
Description
Designed with phlebotomists in mind. Fisherbrand™ Blood Collection Tray was developed after extensive research in clinics and hospitals. This lightweight solution for organizing and transporting tubes and smaller items includes a sturdy plastic tray, 36-well tube rack and disposable inserts.
- Lightweight, washable ABS tray
- Tray features a supply drawer, side pockets, and a ratcheting handle with soft rubber grip that rotates out of the way for easy access
- Tray includes a Heathrow Onerack™ tube rack and two disposable polystyrene inserts
- Additional inserts sold in packs of 4 in two different configurations
Order Info
Additional inserts sold in packs of 4.
Specifications
Blue | |
Blood collection | |
2 inserts, 13 mm test tube rack, supply drawer and side pockets, and ratcheting handle | |
ABS |
429 x 265 x 172 mm | |
13 mm tube rack and Tray Insert (style A) | |
36 (tube rack) | |
Blood Collection Tray |