missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ BloodBloc™ Biohazard Wipes, Liners and Rolls
Shop All Fisherbrand Products
Click to view available options
Product Type:
Biohazard Liner
Biohazard Roll
Biohazard Wipe
Biohazard Wipes
Absorbancy:
Heavy
Standard
Super
Dimensions (L x W):
100 ft. x 16 in.
16 x 16 in.
22 x 16 in.
3 x 3 in. (7.62 x 7.62 cm)
4 x 4 in. (10.16 x 10.16 cm)
9 x 8 in. (22.86 x 20.32 cm)
Packungsgröße:
Each
Pack of 100
Pack of 125
Pack of 200
Pack of 250
Pack of 50
Beschreibung
Wipes and Liners are non-sterile and not intended for use on patients.
- Soft, white absorbent wipe with bright-orange polyethylene backing is impermeable to blood, body fluids and strong solvents including xylene, xylene substitutes, acetones, ketones, alcohols and formalin
- Nonwoven cellulose captures and retains fluid; nonlinting for use with delicate instrumentation such as blood counters
- Orange flexible impermeable plastic backing prevents soak-through of pathogens and chemicals to gloves which help minimize cross-contamination when lab personal open doors or touch surfaces
- Can also be used as mats and liners; rolls can be cut to desired length
- One time use only
Spezifikation
Spezifikation
| Product Type | Biohazard Wipe |
| Length (English) | 3 in. |
| Length (Metric) | 7.62 cm |
| Width (English) | 3 in. |
| Width (Metric) | 7.62 cm |
| Material | Non-woven Plastic Cellulose |
| Absorbancy | Standard |
| Color | White, Orange |
| Disposable | Single-use |
| Dimensions (L x W) | 3 x 3 in. (7.62 x 7.62 cm) |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur