Learn More
Fisherbrand™ Borosilicate Glass Disposable Serological Pipets with Regular Tip, Standard Length, Nonsterile
Long, slender, tapered tips speed pipetting
492.76 €
Specifications
Certifications/Compliance USP type I, ASTM E438
Material Borosilicate Glass
Sterility Non-sterile
Graduated Yes
Tip Style Regular
View More Specs
Products 1
Product Code Brand Volume (Metric) Graduations Negative Graduations Color Price Quantity & Availability  
Product Code Brand Volume (Metric) Graduations Negative Graduations Color Price Quantity & Availability  
13267499
View Documents
Fisherbrand™
72120-11100
1 mL 0.01 mL 0.2 mL Yellow This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Regular-tipped pipets have long, slender, tapered tips to speed pipetting
Chemically inert pipets have negative graduations as well as bidirectional scales.
  • Blue markings are easy to read
  • Packaging meets requirements of small-, medium-, or large-volume use
  • Multipack with resealable flap for larger volume use permits easy access to several pipets at a time without contaminating unused pipets
  • Outer packaging is recyclable
Order Info
Bulk packed.
Compliance
Regular-tip styles have volume tolerances that comply with ASTM™ E 714. Calibrated To Deliver/Blow Out,
Specifications
Specifications
USP type I, ASTM E438
Non-sterile
Regular
Serological Pipet
Borosilicate Glass
Yes
Bulk
Single-use