missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Cell Spreaders

Nonbreakable and reusable polypropylene spreaders available in two sizes: 30 and 60mm bar widths

Brand:  Fisherbrand™ 0810012

16.28 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.


 View more versions of this product

Product Code. 15635467

  • 20.00 € / Pack of 25
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Nonbreakable and reusable polypropylene spreaders available in two sizes: 30 and 60mm bar widths

  • Use 30mm wide spreaders for prepared biplates; 60mm for 100 to 150mm prepared plates
  • Autoclavable

Order Info

Available sterile (individually wrapped) or nonsterile

Specifications

Specifications

Autoclavable
Prepared Biplates
3 cm
Cell Spreader
Blue
1.18 in.
Polypropylene
Sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.