Learn More
Hellma Cylindrical Cells
Fit standard cell holders
Brand:  Hellma 120-10-40
 View more versions of this product
Product Code. 12711705
  • 221.00 € / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Cylindrical cells have a filler neck and tapered PTFE stopper (two necks and stoppers on 50 and 100mm lightpath cells).
  • Uniform diameter of 22mm
  • Available with Suprasil™ quartz or near-UV glass windows
Specifications
Specifications
Suprasil
Suprasil quartz window; One stopper
10 mm