Learn More
Fisherbrand™ Digital Orbital Shaker
Packed full of high-end design features, making it a versatile shaker for a vast range of protocols. Shaker has a speed range of 20-300 rpm in 1 rpm increments.
Brand:  Fisherbrand™ 120905
844.84 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Product Code. 17811682
  • 1012.96 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • 5 Year Warranty
  • No-stop electronic motor feedback enables the unit to instantly adjust to weight load changes and unbalanced loads for truly reproducible results
  • Suitable for use in incubators (excl. CO2) and cold rooms within the stated temperature range
  • Soft-start and shut down feature at higher speeds reduces spill instances from full vessels
  • A high weight loading of 4.5 kg (10 lb.), with an overload protection cut out
  • Universal hi-lo platform and bungee cords allows you to create your own holding matrix for vessels of all shapes and sizes
  • Back lit display is easy to read.
  • Time and speed can be separately set
  • Very low start speed for sensitive sample agitation, with a high top end and good stability for more vigorous applications
  • Powered by 12 VDC, includes double insulated worldwide power adapter, 110 V to 240 V, supplied with 4 different plugs for power adapter
  • Long-life maintenance free motor and drive
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Digital Orbital Shaker > Product Type: Orbital platform shaker