Learn More
Fisherbrand™ Disposable Borosilicate Glass Tubes with Threaded End
Available with or without marking spot
Brand:  Fisherbrand™ FS73750-16150
 View more versions of this product
Product Code. 11537423
  • 468.00 € / Pack of 1000
Estimated Shipment: 02-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Available with or without marking spot
  • Round bottom
  • Without caps
Order Info
Supplied in shrink-wrapped shelf trays of 500
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ASTM E438, Type I, Class B
150 mm
Single-use
Marking Spot
15-415
16 mm
Disposable Culture Tube
20 mL
Borosilicate Glass
For tissue culture work and general bacteriological use
Tray
Non-sterile
Not Included
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products