missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Disposable Cuvettes

Product Code. 11682599 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
1.5 mL
4.5 mL
Type:
Cuvette
Semi-micro Cell
Standard Cuvette
Material:
Methacrylate
PMMA
Polystyrene
Wavelength Range:
285 to 750 nm
340 to 750 nm
Unit Size:
Pack of 500

Product Code. 11682599

Only null left
Add to basket
Edge
Add to basket

Choice of polystyrene or methacrylate

Fisherbrand™ Disposable Cuvettes are available in choice of polystyrene or methacrylate. Select polystyrene cuvets for assays in visible spectral range (340 to 750nm); for accuracy throughout UV/Vis range (285 to 750nm), choose cuvets made from a superior grade of methacrylate.

  • Disposable 10mm pathlength cuvets designed for visible and UV/Vis assays
  • Clear four-sided cuvets are ideal for fluorimetry and nephelometry, as well as spectrophotometry
  • Scale down assay volumes and save on reagents by using semimicro cuvets with capacity to 1.5mL
  • Molded-in arrow indicates direction of transmission on standard and semimicro cuvets—helping assure uniformity
  • Each case contains cuvets with same mold impression number—assuring precision between samples—standard deviation is <±0.005 extinction units between cuvets

Order Info

Packaged 100 cuvets per stackable tray; five trays to a case. Outside dimensions: 0.47L x 0.47W x 1.77 in. H (12 x 12 x 45mm).

Specifications

Type Standard Cuvette
Disposable Single-use
Capacity (Metric) 4.5 mL
Material Polystyrene
Length (Metric) 12mm
Quantity 500 Case
Path Length 10 mm
Wavelength Range 340 to 750 nm
Height (Metric) 45 mm
For Use With (Equipment) Spectrophotometers, Fluorometers, Nephelometers
Width (Metric) 12 mm
For Use With (Application) For assays in visible spectral range, accuracy throughout UV/Visible range
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.