missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Disposable Cuvettes
Description
Fisherbrand™ Disposable Cuvettes are available in choice of polystyrene or methacrylate. Select polystyrene cuvets for assays in visible spectral range (340 to 750nm); for accuracy throughout UV/Vis range (285 to 750nm), choose cuvets made from a superior grade of methacrylate.
- Disposable 10mm pathlength cuvets designed for visible and UV/Vis assays
- Clear four-sided cuvets are ideal for fluorimetry and nephelometry, as well as spectrophotometry
- Scale down assay volumes and save on reagents by using semimicro cuvets with capacity to 1.5mL
- Molded-in arrow indicates direction of transmission on standard and semimicro cuvets—helping assure uniformity
- Each case contains cuvets with same mold impression number—assuring precision between samples—standard deviation is <±0.005 extinction units between cuvets
Order Info
Packaged 100 cuvets per stackable tray; five trays to a case. Outside dimensions: 0.47L x 0.47W x 1.77 in. H (12 x 12 x 45mm).
Specifications
Semi-micro Cell | |
1.5 mL | |
12mm | |
10 mm | |
45 mm | |
12 mm |
Single-use | |
Methacrylate | |
500 Case | |
285 to 750 nm | |
Spectrophotometers, Fluorometers and Nephelometers | |
For assays in visible spectral range, accuracy throughout UV/Visible range |
Fisherbrand Focus on glassware and plasticware
File size: 66.4 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Disposable Cuvettes > Semimicro; Methacrylate
Spot an opportunity for improvement?Share a Content Correction