missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Disposable Cuvettes

Choice of polystyrene or methacrylate

Brand:  Fisherbrand™ 14955129

 View more versions of this product

Product Code. 11954395

  • 389.00 € / Pack of 500
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Fisherbrand™ Disposable Cuvettes are available in choice of polystyrene or methacrylate. Select polystyrene cuvets for assays in visible spectral range (340 to 750nm); for accuracy throughout UV/Vis range (285 to 750nm), choose cuvets made from a superior grade of methacrylate.

  • Disposable 10mm pathlength cuvets designed for visible and UV/Vis assays
  • Clear four-sided cuvets are ideal for fluorimetry and nephelometry, as well as spectrophotometry
  • Scale down assay volumes and save on reagents by using semimicro cuvets with capacity to 1.5mL
  • Molded-in arrow indicates direction of transmission on standard and semimicro cuvets—helping assure uniformity
  • Each case contains cuvets with same mold impression number—assuring precision between samples—standard deviation is <±0.005 extinction units between cuvets

Order Info

Packaged 100 cuvets per stackable tray; five trays to a case. Outside dimensions: 0.47L x 0.47W x 1.77 in. H (12 x 12 x 45mm).

Specifications

Specifications

Cuvette
Single-use
Polystyrene
500 Case
340 to 750 nm
Spectrophotometers, Fluorometers and Nephelometers
For assays in visible spectral range, accuracy throughout UV/Visible range
Four-clear-sided
4.5 mL
12mm
10 mm
45 mm
12 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers