missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Disposable Divided Well Pipette Basins
Disposable basins offer convenience, fast cleanup
75.70 € - 96.82 €
Product Code | Brand | Sterility | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sterility | Packaging | Price | Quantity & Availability | |||||
16666015
|
Fisherbrand™
13681505 |
Sterile | Individually Wrapped |
96.82 €
Pack of 50 |
In Stock
Log in to see stock. ![]() |
|||||
15671424
|
Fisherbrand™
13681506 |
Sterile | 10/Bag |
75.70 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||||
Description
Disposable basins offer convenience, fast cleanup
- Easy-pour corners for quick disposal of excess sample solution
- Packaged in easy-to-open bags to reduce contamination risks
- Divided well design allows for the pipetting of two reagents at the same time
- Dimensions: 5.7L × 2.4W × 0.8 in.H (14.5 × 6.3 × 2.1cm)
Specifications
25 mL | |
Polystyrene |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Disposable Divided Well Pipette Basins