missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Disposable Petri Dishes
Tight-fitting lid prevents dehydration.
101.97 €
Specifications
Disposable | Single-use |
---|---|
Material | Polystyrene |
Sterility | Sterile |
Diameter (Metric) | 47 mm |
Description
Secure, tight-fitting lid helps to prevent dehydration.
Available with or without absorbant pads
Sterilized Petri Dishes
- 47mm dishes can be used with solid agar medium or add absorbent pads for liquid medium
- Molded polystyrene
- Preloaded 47mm dishes with sterilized absorbent pads
- Minimize contamination
- Pure cellulose pad
- Extended 3-year shelf-life
Specifications
Single-use | |
Polystyrene | |
Sterile | |
47 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Disposable Petri Dishes