Learn More
Fisherbrand™ Cryogenic Dots in Laser Sheets
GREENER_CHOICE
Intended uses of cryostorage with liquid nitrogen, refrigeration and ultra-low temperature freezer applications. Come in an easy to use dispenser box with 20 sheets per pack.
Brand:  Fisherbrand™ 11010207
 View more versions of this product
Product Code. 17897191
  • 73.00 € / 25 sheets
Estimated Shipment: 11-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The labels can be printed on and will not bleed. Perfect for distinguishing amongst various specimens.
Specifications
Specifications
Assorted
Non-sterile
0.37 in.
Dot label laser sheet
0.5 to 1.5mL tubes
9.55 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Cryogenic Dots in Laser Sheets > Assorted