Learn More
Fisherbrand™ Elite Pipette Kit
GREENER_CHOICE
Kit provides four adjustable-volume pipettes and a stand.
Brand:  Fisherbrand™ 4680100
796.65 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
Product Code. 15268638
  • 1050.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Kit provides four adjustable-volume pipettes and a stand.
  • Four popular pipette sizes (0.2-2μL, 2-20μL, 20-200μL, 100-1000μL) and stand
  • Compatible with universal fit pipet tips
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1
Pipette Kit
Four pipette sizes (0.2-2uL, 2-20uL, 20-200uL, 100-1000uL) and stand
Elite
0.2-2 μL, 2-20 μL, 20-200 μL, 100-1000 μL
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Elite Pipette Kit > Pipette Kit