missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Fluorescence Rectangular Cells: Cell for Magic Stirrer-Semi-Micro With PTFE Lids, 4 Clear Windows
Fit standard spectrophotometer cell holders
Brand: Fisherbrand™ 14958109
Includes: PTFE lid or stopper
Description
- All cells are 12.5mm wide
- 1, 5 or 10mm path length rectangular cells
- Reduce reagent use with micro cells
- Wavelength range for all cells is 200 to 2500nm
- Quartz windows for transmission from far ultraviolet through the infrared
Specifications
Semi-micro Fluorescence Cell | |
Quartz | |
4mm | |
10 mm | |
49.5 mm | |
12.5 mm | |
Rectangle |
1500 μL | |
PTFE | |
Ea., 4 Ea./Cs. | |
200 to 2500 nm | |
Standard spectrophotometer cell holder | |
Transmission from far ultraviolet through the infrared |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction