missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Fluorescence Rectangular Cells: Macro Cells With PTFE Lids
Includes: PTFE lid or stopper
Description
Fit standard spectrophotometer cell holders
- All cells are 12.5mm wide
- 10mm path length rectangular cells
- Reduce reagent use with micro cells
- Wavelength range for all cells is 200 to 2500nm
- Quartz windows for transmission from far ultraviolet through the infrared
Specifications
Macro Fluorescence Cell | |
Quartz | |
10mm | |
10 mm | |
45 mm | |
12.5 mm | |
Rectangle |
3000 μL | |
PTFE | |
4/Pk., 4 Pk./Cs. | |
200 to 2500 nm | |
Standard Spectrophotometer Cell Holder | |
Transmission from far ultraviolet through the infrared |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction