missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Fluorescence Rectangular Cells: Micro Cells With PTFE Lids

Product Code. 15266003 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
12 μL
45 μL
100 μL
Path Length:
1.5 mm
3 mm
10 mm
Description:
Center H: 15mm
Center H: 8.5mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15266003

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

PTFE lid or stopper

Fit standard spectrophotometer cell holders

  • All cells are 12.5mm wide
  • 1, 5 or 10mm path length rectangular cells
  • Reduce reagent use with micro cells
  • Wavelength range for all cells is 200 to 2500nm
  • Quartz windows for transmission from far ultraviolet through the infrared

Specifications

Type Micro Fluorescence Cell
Description Center H: 8.5mm
Capacity (Metric) 12 μL
Material Quartz
Closure Material PTFE
Width (Metric) Interior 1.5mm
Quantity Ea., 2 Ea./Cs.
Path Length 1.5 mm
Wavelength Range 200 to 2500 nm
Height (Metric) 45 mm
For Use With (Equipment) Standard Spectrophotometer Cell Holder
Width (Metric) 12.5 mm
For Use With (Application) Transmission from far ultraviolet through the infrared
Shape Rectangle
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.