missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Gel-Loading Aerosol-Barrier Tips
GREENER_CHOICE

Help minimize liquid retention

189.52 €

Specifications

Filtered Filtered
Color Natural
For Use With (Application) Help minimize liquid retention
Tip Style Gel Loading
Material Polypropylene
View More Specs

Products 2
Product Code Brand Volume (Metric) Packaging Price Quantity & Availability  
Product Code Brand Volume (Metric) Packaging Price Quantity & Availability  
12992571
GREENER_CHOICE
View Documents
Fisherbrand™
F010R204S20-FIS
20 μL Racked
189.52 €
Pack of 1020
In Stock
to see stock.
Edge
Add to basket
 
11967734
GREENER_CHOICE
View Documents
Fisherbrand™
F010R204S100FIS
10 to 100 μL Racked
189.52 €
Pack of 1020
In Stock
to see stock.
Add to basket
 
Description

Description

  • No RNase or DNase contamination
  • Natural color
  • For use with, but not limited to: Biohit Proline™ Electronic, BrandTech™ Transferpette™, Eppendorf™ Reference 2000, Eppendorf Research 2100, Thermo Finnpipette™ Focus, Fisherbrand™ Finnpipette™, Fisherbrand Finnpipette II, Genemate™ ISC Bio Express, Hamilton SoftGrip™, Jencons Sealpette™ Electronic, Labnet Labpette™ Digital, Mettler Toledo VoluMate™, Nichiryo 5000, Oxford™ Benchmate™, Pipetman™, Rainin™, Vistalab Ovation

Specifications

Specifications

Filtered
Help minimize liquid retention
Polypropylene
Natural
Gel Loading
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Gel-Loading Aerosol-Barrier Tips

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.