missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ HDPE Liquid Samplers

For industrial and environmental applications.

Brand:  Fisherbrand™ 033382B

 View more versions of this product

Product Code. 16562440

  • 247.00 € / Each
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

Chemical-resistant, high-density polyethylene samplers ideal for use in corrosive environments and hard-to-reach places.

  • 500mL bottle screws into the end of handle and is filled via four ports
  • Handle is supplied in two pieces
  • Reusable
  • Handle O.D: 1.25 in. (3.2cm)

Order Info

Additional liquid sampler sizes available. To order, contact your Fisher Scientific Customer Service Representative.

Specifications

Specifications

Liquid Sampler
For industrial and environmental applications
3.2 cm
205.89 cm
1.8 m
7.62 cm
HDPE
1.25 in.
81.06 in.
6 ft.
3 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers