Learn More
Fisherbrand™ Heavy-Duty/Utility Funnels
Ideal for continuous lab work and heavy-duty jobs
27.70 € - 283.00 €
Product Code | Brand | Capacity (Metric) | Diameter (Metric)Top | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric)Top | Height (Metric) | Price | Quantity & Availability | |||
11997884
|
Fisherbrand™
300800-0800 |
500 mL | 20.3 cm | 20.5 cm | N/A | ||||
12622445
|
Fisherbrand™
300800-0800 |
500 mL | 20.3 cm | 20.5 cm |
27.70 €
Pack of 3 |
In Stock
Log in to see stock. |
|||
11997934
|
Fisherbrand™
300800-0234 |
40 mL | 6.5cm | 9 cm |
142.00 €
Pack of 72 |
Estimated Shipment: 10-04-2025
Log in to see stock. |
|||
12662435
|
Fisherbrand™
300800-0234 |
40 mL | 6.5cm | 9 cm | N/A | ||||
12682435
|
Fisherbrand™
300800-0314 |
100 mL | 8.7cm | 11 cm | N/A | ||||
11907944
|
Fisherbrand™
300800-0314 |
100 mL | 8.7cm | 11 cm |
174.00 €
Pack of 72 |
Estimated Shipment: 10-04-2025
Log in to see stock. |
|||
12602445
|
Fisherbrand™
300800-0638 |
410 mL | 15.5cm | 17.7 cm | N/A | ||||
11917914
|
Fisherbrand™
300800-0638 |
410 mL | 15.5cm | 17.7 cm |
202.00 €
Pack of 24 |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||
12692435
|
Fisherbrand™
300800-0500 |
210 mL | 12.5cm | 14.1 cm | N/A | ||||
11927944
|
Fisherbrand™
300800-0500 |
210 mL | 12.5cm | 14.1 cm |
283.00 €
Pack of 72 |
Estimated Shipment: 10-04-2025
Log in to see stock. |
|||
12672435
|
Fisherbrand™
300800-0414 |
140 mL | 10.3cm | 11.5 cm | N/A | ||||
11917944
|
Fisherbrand™
300800-0414 |
140 mL | 10.3cm | 11.5 cm |
162.00 €
Pack of 72 |
Estimated Shipment: 10-03-2025
Log in to see stock. |
|||
Description
Ideal for continuous lab work and heavy-duty jobs
- Durable polypropylene construction with external ridges and ribbing to prevent vapor lock
- Autoclavable to 120°C (248°F)
Specifications
Polypropylene | |
Autoclavable |