missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ High-Form Porcelain Crucibles

Withstand temperatures up to 1150°C (2102°F)

Brand:  Fisherbrand™ FB965F

 View more versions of this product

Product Code. 13409709

  • 32.15 € / Pack of 6
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Alert:

Alert: Heating and cooling rates should not exceed 200°C per hour and maximum use temp of 1150°C


Description

Description

Withstand temperatures up to 1150°C (2102°F)

  • Glazed inside and out except for outside bottom surface and rim

Order Info

Covers available separately. See Fisherbrand™ Porcelain Crucible Covers (FB965R through FB965Z).

Specifications

Specifications

Porcelain
1150°C
32g
50mm
Crucible
Glazed
2 mm
40 mL
40mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers