missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Lab Hazard Warning Label Refills

Printed in safety colors.

2.00 €

Specifications

Color Assorted
Format Tape
For Use With (Application) Printed in safety colors
Length (English) 57 yd.
Length (Metric) 20.3 m
View More Specs

Products 1
Product Code Brand Display Text Price Quantity & Availability  
Product Code Brand Display Text Price Quantity & Availability  
17477282
View Documents
Fisherbrand™
15-933
Corrosive
2.00 €
1 roll
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Printed in safety colors.

For use with Fisherbrand™ Lab Hazard Warning Label Dispenser (cat. no. 15931).

  • Tape rolls preprinted with warnings in safety colors: POISON (yellow on black); CORROSIVE (black on yellow); FLAMMABLE (red on white); and RADIOACTIVE (red on yellow)
  • Serrated tear-off edge for easy tape separation
  • Approximately 200 imprints per roll

Order Info

Each tape roll measures 0.50 in. W x 22 yd. L (13mm x 20m)

Specifications

Specifications

Assorted
Printed in safety colors
20.3 m
Tape
57 yd.
0.5 in.
Videos
SDS
Documents

Documents

Product Certifications