missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Locking-Lid Microcentrifuge Tubes with Polypropylene Snap-Cap™

Product Code. 11956955 Shop All Fisherbrand Products
Click to view available options
Color:
Assorted
Natural
Capacity (Metric):
0.6 mL
1.5 mL
2 mL
Max. RCF:
25,000 x g
26,000 x g
30,000 x g
Packaging:
Bag
Unit Size:
Pack of 1000
Pack of 500

Product Code. 11956955

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Convenient, pierceable locking lids snap into place with one-handed operation

  • Polypropylene tubes with flat tops
  • Ideal for sample-handling applications, including centrifugation, boiling and freezing of specimens, incubation and storage
  • With graduation marks for volume estimation
  • RNase and DNase free

Centrifuges that develop excessive heat may damage this tube.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RNase, DNase-free
Closure Material Polypropylene
Autoclavable Autoclavable
Capacity (Metric) 2 mL
Color Natural
Material Polypropylene
For Use With (Application) Ideal for sample-handling applications, including centrifugation, boiling and freezing of specimens, incubation and storage
Packaging Bag
Closure Type Snap Cap
Max. RCF 25,000 x g
Graduations 0.1 mL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.