missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Microprobes

Six stainless-steel probes for electronics, dissecting, inspecting, soldering, gluing, and micromanipulation

Brand:  Fisherbrand™ 3310

Product Code. 11744356

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Six stainless-steel probes for electronics, dissecting, inspecting, soldering, gluing, and micromanipulation

Excellent for hooking small springs, packing columns, cleaning miniature parts, and soldering boards. Solder would not stick to probes.

  • Tips are hand-honed for sharpness
  • Precision-ground, highly polished, and tempered for long life

Order Info

Pack includes one each of six shapes shown in photo

Specifications

Specifications

Microprobe
Ideal for lab applications like positioning samples under a microscope, dissecting, inspecting, soldering, gluing, and all micro-manipulation in the lab
Needle
15.2 cm
Stainless Steel
Six stainless steel probes, carrying case
6 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Microprobes > Length: 6 in.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.