missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Extremely durable and economically priced holder for standard microscope slides
- Capacity for twenty-five (25) 3 x 1 in. or 75 x 25mm standard slides
- Tight-fitting lid protects slides and prevents dust accumulation
- Durable molded black plastic construction
- Preprinted index card in lid for easy slide identification
- Front has tab area (tab included) for additional identification
- Base has interlocking grooves for stacking several units
- Slides stay securely in place with numbered slide partitions
Specifications
25 | |
Black | |
Numbered slide partitions | |
75 mm | |
1 in. |
Snap-on | |
Microscope Slide | |
3 in. | |
Plastic | |
25 mm |