missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Nichrome/Aluminum Inoculating Loops

Fisherbrand Inoculating Loops consist of a nichrome wire loop mounted to an aluminum handle. Inoculating loops are used to introduce microorganisms into a culture medium.

Brand:  Fisherbrand™ 131045

Product Code. 16002282

  • 24.00 € / Pack of 12
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Fisherbrand Nichrome/Aluminum Inoculating Loops consist of a nichrome wire loop mounted to an aluminum handle. Inoculating loops are used to introduce microorganisms into a culture medium.

  • Fisherbrand™ Nichrome/Aluminum Inoculating Loops are provided with a standard aluminum handle
  • The aluminum handle is available in either an insulated or non-insulated format
  • Inoculating loops are available in a variety of gauges, loop-hole sizes, and wire types to accommodate an array of laboratory requirements.
  • Ideal for selecting, inoculating, or transferring microbiological samples
  • Loops slide along agar surfaces without cutting or tearing material/sample
  • Cools rapidly after sterilization
  • Handle length: 8in.(203mm)
  • Passivated
  • Autoclavable up to 135°C; Can be sterilized by Eto/gamma radiation
  • Latex-free
  • Produced under ISO 9002, GMP, cGMP guidelines
Specifications

Specifications

3.93 in.
Nichrome wire loop; Aluminum handle
Silver
25 G
8 in.
10 cm
Sterile
Inoculating Loop
Tapered end aluminum handle
20.3 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.