missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Nickel Bow Tongs

Designed using high quality nickel 200 components these tongs have good mechanical properties and excellent resistance to many corrosive environments.

Brand:  Fisherbrand™ 3050

 View more versions of this product

Product Code. 11562692

  • 28.20 € / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Laboratory Bow Tong, nickel 200 spec (minimum 99.6% nickel content), bright finish
  • Made of Nickel 200 that is commercially pure (99.6%) wrought nickel
  • Has good mechanical properties and excellent resistance to many corrosive environments
  • Features high thermal and electrical conductivities, low gas content and low vapor pressure
Specifications

Specifications

55 mm
152 mm
Laboratory Bow Tongs
1 Ea.
Nickel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers