missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ PETG Media Bottles PROMO
![GREENER_CHOICE](/content/dam/fishersci/glyphs/green_glyph.png)
Click to view available options
Capacity (Metric):
1000 mL
Specifications
Specifications
Capacity (Metric) | 1000 mL |
Color | Clear |
For Use With (Application) | For media preparation and storage |
Material | PETG |
Quantity | 12 Cs. |
Closure Material | HDPE |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction