Learn More
Fisherbrand™ Plastic Joint Clips
Made of plastic for use on standard taper glass joints
Brand: Fisherbrand™ 14-955-256
This item is not returnable.
View return policy
Description
Made of plastic for use on standard taper glass joints
- Color coded for easy size identification
- Does not corrode
- Will not scratch glass
- Withstands temperatures up to 150°C
Specifications
10/Pk. | |
Plastic | |
Red |
Joint Clip | |
Standard taper Glass joint size 29 |
Product Content Correction
Product Title
Fisherbrand™ Plastic Joint Clips > Red; For joint size: 29
Spot an opportunity for improvement?Share a Content Correction