Learn More
Fisherbrand™ Autoclavable HD Biohazard Waste Bags
Strong and leak-resistant
Brand:  Fisherbrand™ 22044563
 View more versions of this product
Product Code. 18336513
  • 113.00 € / Pack of 200
Explore more special offers
Description
Description
Strong and leak-resistant
  • Printed with biohazard symbol and text in English and Spanish
  • Supplied with twist tie closure
Specifications
Specifications
Twist Tie
23 x 19 in. (58 x 48 cm)
48 cm
Biohazard symbol and text in English and Spanish
23 in.
Polyethylene
Red with Black Print
58 cm
Autoclavable
19 in.
1.25 mil