Learn More
Fisherbrand™ Autoclavable HD Biohazard Waste Bags
Strong and leak-resistant
Brand:  Fisherbrand™ 22044559
 View more versions of this product
Product Code. 18356513
  • 110.00 € / Pack of 100
Explore more special offers
Description
Description
Strong and leak-resistant
  • Printed with biohazard symbol and text in English and Spanish
  • Supplied with twist tie closure
Specifications
Specifications
Twist Tie
30 x 24 in. (76 x 61 cm)
61 cm
Biohazard symbol and text in English and Spanish
30 in.
Polyethylene
Clear with Red Print
76 cm
Autoclavable
24 in.
1.50 mil