Learn More
Fisherbrand™ Autoclavable HD Biohazard Waste Bags
Strong and leak-resistant
Brand:  Fisherbrand™ 22044558
 View more versions of this product
Product Code. 18568056
  • 51.00 € / Pack of 100
Explore more special offers
Description
Description
Strong and leak-resistant
  • Printed with biohazard symbol and text in English and Spanish
  • Supplied with twist tie closure
Specifications
Specifications
Twist Tie
24 x 12 in. (61 x 30 cm)
30 cm
Biohazard symbol and text in English and Spanish
24 in.
Polyethylene
Clear with Red Print
61 cm
Autoclavable
12 in.
1.50 mil