missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Porcelain Crucible Covers

For use with Fisherbrand High- and Wide-Form Porcelain Crucibles.

Brand:  Fisherbrand™ FB965Z

 View more versions of this product

Product Code. 13499709

  • 54.65 € / Pack of 6
Estimated Shipment: 10-06-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

For use with Fisherbrand High- and Wide-Form Porcelain Crucibles

  • Glazed inside and out except for the outside bottom surface
  • With perforated handle
  • Strongly recommended that heating and cooling rates not exceed 200°C per hour and maximum use temperature of 1100°C

Specifications

Specifications

With Perforated handle
Porcelain
Use with Fisherbrand high and wide form porcelain crucible
3mm
115mm
Crucible Cover
105mm
Glazed
1150°C
119g
9mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Porcelain Crucible Covers > 115 x 9 x 3mm; Inner Dia.: 105mm; 119g; Case wt.: 7.35kg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.