missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Porcelain Evaporating Dishes

Glazed inside and out except for rim.

Brand:  Fisherbrand™ FB968F

 View more versions of this product

Product Code. 13439729

  • 157.00 € / Pack of 6
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

Porcelain dish with lip

  • Round bottom
  • Glazed inside and out except for rim.
  • Highly resistant to chemicals
  • Withstand temperatures up to 1150°C (2102°F)
  • Without cover

Specifications

Specifications

Round
Porcelain
Evaporating Dish
Glazed
45 mm
4.52 in.
250 mL
Round
Evaporating Application
3 mm
1.77 in.
115 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers